Product Details
- SNP ID
-
rs56402864
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
15
- Location
-
Chr.1:2184675 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGACACACAGTTCACCAGCGAGCC[C/G]GTGCAGCTGACCCCAGACGATGAGT
- Phenotype
-
MIM: 615183
MIM: 176982
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FAAP20
PubMed Links
Gene Details
- Gene
- FAAP20
- Gene Name
- Fanconi anemia core complex associated protein 20
- Gene
- LOC100506504
- Gene Name
- uncharacterized LOC100506504
- Gene
- PRKCZ
- Gene Name
- protein kinase C zeta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001033581.1 |
1737 |
Silent Mutation |
CCC,CCG |
P373P |
NP_001028753.1 |
NM_001033582.1 |
1737 |
Silent Mutation |
CCC,CCG |
P373P |
NP_001028754.1 |
NM_001242874.1 |
1737 |
Silent Mutation |
CCC,CCG |
P452P |
NP_001229803.1 |
NM_002744.4 |
1737 |
Silent Mutation |
CCC,CCG |
P556P |
NP_002735.3 |
XM_011541773.1 |
1737 |
Intron |
|
|
XP_011540075.1 |
XM_011541774.1 |
1737 |
Intron |
|
|
XP_011540076.1 |
XM_011541775.2 |
1737 |
Intron |
|
|
XP_011540077.1 |
XM_011541776.1 |
1737 |
Intron |
|
|
XP_011540078.1 |
XM_011541778.1 |
1737 |
Intron |
|
|
XP_011540080.1 |
XM_017001789.1 |
1737 |
Intron |
|
|
XP_016857278.1 |
XM_017001790.1 |
1737 |
Silent Mutation |
CCC,CCG |
P564P |
XP_016857279.1 |
XM_017001791.1 |
1737 |
Silent Mutation |
CCC,CCG |
P474P |
XP_016857280.1 |
XM_017001792.1 |
1737 |
Intron |
|
|
XP_016857281.1 |
XM_017001793.1 |
1737 |
Intron |
|
|
XP_016857282.1 |
XM_017001794.1 |
1737 |
Intron |
|
|
XP_016857283.1 |
XM_017001795.1 |
1737 |
Intron |
|
|
XP_016857284.1 |
XM_017001796.1 |
1737 |
Silent Mutation |
CCC,CCG |
P381P |
XP_016857285.1 |
XM_017001797.1 |
1737 |
Intron |
|
|
XP_016857286.1 |
XM_017001798.1 |
1737 |
Silent Mutation |
CCC,CCG |
P381P |
XP_016857287.1 |
XM_017001799.1 |
1737 |
Silent Mutation |
CCC,CCG |
P373P |
XP_016857288.1 |
XM_017001800.1 |
1737 |
Intron |
|
|
XP_016857289.1 |
XM_017001801.1 |
1737 |
Silent Mutation |
CCC,CCG |
P369P |
XP_016857290.1 |
XM_017001802.1 |
1737 |
Intron |
|
|
XP_016857291.1 |
XM_017001803.1 |
1737 |
Intron |
|
|
XP_016857292.1 |
XM_017001804.1 |
1737 |
Silent Mutation |
CCC,CCG |
P319P |
XP_016857293.1 |
XM_017001805.1 |
1737 |
Silent Mutation |
CCC,CCG |
P319P |
XP_016857294.1 |
View Full Product Details