Product Details

SNP ID
rs56280055
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:161318291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTCCCCCTCCCCCGCATATAAA[C/T]ATGTTTATGCTTGGCTAATTAAGAA
Phenotype
MIM: 159440 MIM: 602413
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPZ PubMed Links

Gene Details

Gene
MPZ
Gene Name
myelin protein zero
There are no transcripts associated with this gene.

Gene
SDHC
Gene Name
succinate dehydrogenase complex subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035511.1 Intron NP_001030588.1
NM_001035512.1 Intron NP_001030589.1
NM_001035513.1 Intron NP_001030590.1
NM_001278172.1 Intron NP_001265101.1
NM_003001.3 Intron NP_002992.1

View Full Product Details