Product Details

SNP ID
rs56391640
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:203216770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGAGCAGAAAGCCTGGATAAAG[A/G]AAGACCACAGAAAGGCCTGCAGGAG
Phenotype
MIM: 600031
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHIT1 PubMed Links

Gene Details

Gene
CHIT1
Gene Name
chitinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256125.1 1559 UTR 3 NP_001243054.2
NM_001270509.1 1559 UTR 3 NP_001257438.1
NM_003465.2 1559 UTR 3 NP_003456.1

View Full Product Details