Product Details

SNP ID
rs61749335
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:223784160 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATCCCTCCTTATCATTAAGGCG[C/T]GCCCACCACCATTCGATTTCATCTT
Phenotype
MIM: 114230 MIM: 602143
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAPN2 PubMed Links

Gene Details

Gene
CAPN2
Gene Name
calpain 2
There are no transcripts associated with this gene.

Gene
TP53BP2
Gene Name
tumor protein p53 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031685.2 3610 Silent Mutation GCA,GCG A1106A NP_001026855.2
NM_005426.2 3610 Silent Mutation GCA,GCG A977A NP_005417.1
XM_011544268.1 3610 Silent Mutation GCA,GCG A1039A XP_011542570.1
XM_011544269.2 3610 Silent Mutation GCA,GCG A977A XP_011542571.1
XM_017002220.1 3610 Silent Mutation GCA,GCG A1039A XP_016857709.1

View Full Product Details