Product Details
- SNP ID
-
rs58901374
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
14
- Location
-
Chr.1:235978371 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGACAGAAGAAGATGAGGGAAGCT[A/G]AGAAGGAAAGTGGGTGAGGGGAGAG
- Phenotype
-
MIM: 131390
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NID1
PubMed Links
Gene Details
- Gene
- NID1
- Gene Name
- nidogen 1
View Full Product Details