Product Details

SNP ID
hCV88461735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223053277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGGCAGACGATGAGCTGGAGGA[G/T]ACCTCGGAGACGCCCCTCAACGAGA
Phenotype
MIM: 607775
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNE4 PubMed Links

Gene Details

Gene
KCNE4
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080671.3 815 Missense Mutation GAG,GAT E200D NP_542402.3

View Full Product Details