Product Details

SNP ID
rs60905059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:127638276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGAGTGAAAGAAGAGGAAAAAG[A/G]CCTCCTTTAATTTGTTTTCCTGTCC
Phenotype
MIM: 616627
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PODXL2 PubMed Links

Gene Details

Gene
PODXL2
Gene Name
podocalyxin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015720.3 Intron NP_056535.1

View Full Product Details