Product Details

SNP ID
rs59249742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:136251335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGCTTGCCCAGTGTGTCCGTG[A/G]CTGGTGGGTGAGTCTGGACCGTTGT
Phenotype
MIM: 232050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCCB PubMed Links

Gene Details

Gene
PCCB
Gene Name
propionyl-CoA carboxylase beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000532.4 Intron NP_000523.2
NM_001178014.1 Intron NP_001171485.1
XM_011512873.1 Intron XP_011511175.1

View Full Product Details