Product Details

SNP ID
rs119470018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:158646896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGTTCCGTTTCTAACTTTTATTA[A/G]CAAATTGGACCGAATGGGCTCCAAC
Phenotype
MIM: 606639
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GFM1 PubMed Links

Gene Details

Gene
GFM1
Gene Name
G elongation factor mitochondrial 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308164.1 659 Missense Mutation AAC,AGC N174S NP_001295093.1
NM_001308166.1 659 Missense Mutation AAC,AGC N174S NP_001295095.1
NM_024996.5 659 Missense Mutation AAC,AGC N174S NP_079272.4
XM_006713795.1 659 Missense Mutation AAC,AGC N174S XP_006713858.1

View Full Product Details