Product Details

SNP ID
rs62272783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160498473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATGATGCGTATCACCCCTGTGA[C/T]TGGACTACAATGCAGGGCAAAGGTG
Phenotype
MIM: 602970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KPNA4 PubMed Links

Gene Details

Gene
KPNA4
Gene Name
karyopherin subunit alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002268.4 5503 UTR 3 NP_002259.1

View Full Product Details