Product Details

SNP ID
rs55948310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15343087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTAGAGAATGTTAGCTATTCTTA[C/T]TAGCTATTAAATGCTGATTTCTTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1QTNF7 PubMed Links

Gene Details

Gene
C1QTNF7
Gene Name
C1q and tumor necrosis factor related protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135170.1 Intron NP_001128642.1
NM_001135171.1 Intron NP_001128643.1
NM_031911.4 Intron NP_114117.1
XM_011513772.1 Intron XP_011512074.1
XM_011513773.1 Intron XP_011512075.1
Gene
LOC101929095
Gene Name
uncharacterized LOC101929095
There are no transcripts associated with this gene.

View Full Product Details