Product Details

SNP ID
rs58787312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:37815122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGTATCTTTGCTTTTTTTTTCC[C/T]CTCTCCTTTCCAGGGTATGTCTCCA
Phenotype
MIM: 600837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GDNF PubMed Links

Gene Details

Gene
GDNF
Gene Name
glial cell derived neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000514.3 1385 UTR 3 NP_000505.1
NM_001190468.1 1385 UTR 3 NP_001177397.1
NM_001190469.1 1385 UTR 3 NP_001177398.1
NM_001278098.1 1385 UTR 3 NP_001265027.1
NM_199231.2 1385 UTR 3 NP_954701.1
XM_011514030.2 1385 Intron XP_011512332.1
XM_017009337.1 1385 Intron XP_016864826.1

View Full Product Details