Product Details

SNP ID
hCV88646506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:53484291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTAGCCTATGAGGGAAAGTGTA[A/T]CAGTAGGTATTCTGGATTGAGGAAG
Phenotype
MIM: 136470
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FST PubMed Links

Gene Details

Gene
FST
Gene Name
follistatin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006350.3 1077 Missense Mutation AAC,ATC N240I NP_006341.1
NM_013409.2 1077 Missense Mutation AAC,ATC N240I NP_037541.1

View Full Product Details