Product Details

SNP ID
rs57348345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:14145282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCTCTTTTACAGGGTTTTATTT[A/G]TTTATTTTCTTAAAGTTTACAGCTT
Phenotype
MIM: 604070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGKB PubMed Links

Gene Details

Gene
DGKB
Gene Name
diacylglycerol kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004080.2 6451 UTR 3 NP_004071.1
NM_145695.2 6451 Intron NP_663733.1
XM_005249628.2 6451 Intron XP_005249685.1
XM_005249629.2 6451 Intron XP_005249686.1
XM_005249630.2 6451 Intron XP_005249687.1
XM_005249631.3 6451 Intron XP_005249688.1
XM_011515153.2 6451 Intron XP_011513455.1
XM_011515154.2 6451 Intron XP_011513456.1
XM_011515156.2 6451 Intron XP_011513458.1
XM_011515157.2 6451 Intron XP_011513459.1
XM_011515158.2 6451 Intron XP_011513460.1
XM_017011783.1 6451 Intron XP_016867272.1
XM_017011784.1 6451 Intron XP_016867273.1
XM_017011785.1 6451 Intron XP_016867274.1
XM_017011786.1 6451 Intron XP_016867275.1
XM_017011787.1 6451 Intron XP_016867276.1
XM_017011788.1 6451 Intron XP_016867277.1
XM_017011789.1 6451 Intron XP_016867278.1
XM_017011790.1 6451 Intron XP_016867279.1
XM_017011791.1 6451 Intron XP_016867280.1
XM_017011792.1 6451 Intron XP_016867281.1

View Full Product Details