Product Details

SNP ID
rs62457152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:29566448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCTCCCGCCCAGCCCGCTCCCG[G/T]GGAGCCCACGCCACCTGCGCCGCCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRR15 PubMed Links

Gene Details

Gene
PRR15
Gene Name
proline rich 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175887.2 1634 Missense Mutation GGG,GTG G40V NP_787083.1
XM_005249653.3 1634 Missense Mutation GGG,GTG G40V XP_005249710.1
XM_011515198.1 1634 Missense Mutation GGG,GTG G40V XP_011513500.1
XM_011515199.1 1634 Missense Mutation GGG,GTG G40V XP_011513501.1
XM_017011842.1 1634 Missense Mutation GGG,GTG G40V XP_016867331.1

View Full Product Details