Product Details

SNP ID
hCV88826881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:38384739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTGTAAATCATTAAGAGCATAA[C/T]AACAAAAGTGAACTCTTCAGGTTTT
Phenotype
MIM: 600418
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMPH PubMed Links

Gene Details

Gene
AMPH
Gene Name
amphiphysin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001635.3 2257 UTR 3 NP_001626.1
NM_139316.2 2257 UTR 3 NP_647477.1
Gene
TRG-AS1
Gene Name
T cell receptor gamma locus antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details