Product Details

SNP ID
rs55841161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44072303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATGAGTGGTCTGGGAAATCCTC[A/G]CCTAACAAAGTGGCTTTTGATTCAA
Phenotype
MIM: 612931 MIM: 606344
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6838 PubMed Links

Gene Details

Gene
MIR6838
Gene Name
microRNA 6838
There are no transcripts associated with this gene.

Gene
PGAM2
Gene Name
phosphoglycerate mutase 2
There are no transcripts associated with this gene.

Gene
POLM
Gene Name
DNA polymerase mu
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284330.1 2483 UTR 3 NP_001271259.1
NM_001284331.1 2483 UTR 3 NP_001271260.1
NM_013284.3 2483 UTR 3 NP_037416.1
XM_005249711.2 2483 UTR 3 XP_005249768.1
XM_006715692.2 2483 UTR 3 XP_006715755.1
XM_006715696.2 2483 UTR 3 XP_006715759.1
XM_006715698.3 2483 UTR 3 XP_006715761.1
XM_011515275.1 2483 UTR 3 XP_011513577.1
XM_011515278.1 2483 UTR 3 XP_011513580.1
XM_011515279.1 2483 UTR 3 XP_011513581.1
XM_011515282.2 2483 UTR 3 XP_011513584.1
XM_011515285.1 2483 UTR 3 XP_011513587.1
XM_011515286.2 2483 Intron XP_011513588.1
XM_011515287.2 2483 Intron XP_011513589.1
XM_017011998.1 2483 UTR 3 XP_016867487.1
XM_017011999.1 2483 UTR 3 XP_016867488.1
XM_017012000.1 2483 UTR 3 XP_016867489.1
XM_017012001.1 2483 UTR 3 XP_016867490.1
XM_017012002.1 2483 UTR 3 XP_016867491.1
XM_017012003.1 2483 UTR 3 XP_016867492.1
XM_017012004.1 2483 Intron XP_016867493.1
XM_017012005.1 2483 Intron XP_016867494.1

View Full Product Details