Product Details
- SNP ID
-
hCV88836482
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:5986967 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTACATCAACCTGAGAGGCTGACA[G/T]GTCCTGAGTATTTACTAACTTTTGA
- Phenotype
-
MIM: 600259
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PMS2
PubMed Links
Gene Details
- Gene
- PMS2
- Gene Name
- PMS1 homolog 2, mismatch repair system component
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000535.6 |
1831 |
Missense Mutation |
ATG,CTG |
M600L |
NP_000526.2 |
NM_001322003.1 |
1831 |
Missense Mutation |
ATG,CTG |
M465L |
NP_001308932.1 |
NM_001322004.1 |
1831 |
Missense Mutation |
ATG,CTG |
M465L |
NP_001308933.1 |
NM_001322005.1 |
1831 |
Missense Mutation |
ATG,CTG |
M465L |
NP_001308934.1 |
NM_001322006.1 |
1831 |
Missense Mutation |
ATG,CTG |
M548L |
NP_001308935.1 |
NM_001322007.1 |
1831 |
Missense Mutation |
ATG,CTG |
M494L |
NP_001308936.1 |
NM_001322008.1 |
1831 |
Missense Mutation |
ATG,CTG |
M494L |
NP_001308937.1 |
NM_001322009.1 |
1831 |
Missense Mutation |
ATG,CTG |
M465L |
NP_001308938.1 |
NM_001322010.1 |
1831 |
Missense Mutation |
ATG,CTG |
M413L |
NP_001308939.1 |
NM_001322011.1 |
1831 |
Missense Mutation |
ATG,CTG |
M289L |
NP_001308940.1 |
NM_001322012.1 |
1831 |
Missense Mutation |
ATG,CTG |
M289L |
NP_001308941.1 |
NM_001322013.1 |
1831 |
Missense Mutation |
ATG,CTG |
M409L |
NP_001308942.1 |
NM_001322014.1 |
1831 |
Missense Mutation |
ATG,CTG |
M600L |
NP_001308943.1 |
NM_001322015.1 |
1831 |
Missense Mutation |
ATG,CTG |
M497L |
NP_001308944.1 |
View Full Product Details