Product Details

SNP ID
rs60639203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:138589545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACTATGAACTCAGGACCTGAAA[C/T]GTCCTCAGACAGAGCATCCCCATGG
Phenotype
MIM: 610026
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL22A1 PubMed Links

Gene Details

Gene
COL22A1
Gene Name
collagen type XXII alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152888.2 Intron NP_690848.1
XM_011516883.2 Intron XP_011515185.1
XM_011516884.2 Intron XP_011515186.1
XM_011516885.2 Intron XP_011515187.1
XM_011516886.2 Intron XP_011515188.1
XM_011516887.2 Intron XP_011515189.1
XM_011516888.2 Intron XP_011515190.1
XM_011516889.2 Intron XP_011515191.1
XM_017013150.1 Intron XP_016868639.1
XM_017013151.1 Intron XP_016868640.1
XM_017013152.1 Intron XP_016868641.1

View Full Product Details