Product Details

SNP ID
rs55750068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34458836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTAACCGCGTCAGGGAGTTGGA[A/T]TCTATCCTGCAAGGGCACGGGGACC
Phenotype
MIM: 604366
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAI1 PubMed Links

Gene Details

Gene
DNAI1
Gene Name
dynein axonemal intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281428.1 85 UTR 5 NP_001268357.1
NM_012144.3 85 UTR 5 NP_036276.1
XM_006716758.3 85 Intron XP_006716821.1
XM_011517846.1 85 UTR 5 XP_011516148.1
XM_011517847.2 85 UTR 5 XP_011516149.1
XM_011517848.1 85 UTR 5 XP_011516150.1
XM_011517849.1 85 UTR 5 XP_011516151.1
XM_011517850.2 85 UTR 5 XP_011516152.1
XM_017014625.1 85 UTR 5 XP_016870114.1
Gene
FAM219A
Gene Name
family with sequence similarity 219 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184940.1 85 Intron NP_001171869.1
NM_001184941.1 85 Intron NP_001171870.1
NM_001184942.1 85 Intron NP_001171871.1
NM_001184943.1 85 Intron NP_001171872.1
NM_001184945.1 85 Intron NP_001171874.1
NM_147202.1 85 Intron NP_671735.1
XM_005251402.4 85 Intron XP_005251459.1
XM_011517797.2 85 Intron XP_011516099.1
XM_011517798.2 85 Intron XP_011516100.1
XM_011517799.2 85 Intron XP_011516101.1

View Full Product Details