Product Details

SNP ID
hCV89025287
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120816348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTAATATACTGAACTGTACACT[C/T]AAAATAGTTATGACAAATTTTATTG
Phenotype
MIM: 604452
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PSMD5 PubMed Links

Gene Details

Gene
PSMD5
Gene Name
proteasome 26S subunit, non-ATPase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270427.1 3185 UTR 3 NP_001257356.1
NM_005047.3 3185 UTR 3 NP_005038.1

View Full Product Details