Product Details

SNP ID
rs55782000
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127922384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTATACGTGTACAATACATACTCA[A/G]TTTCAAAGACAGGATGAAAAAAAGG
Phenotype
MIM: 612865 MIM: 606378
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PIP5KL1 PubMed Links
Additional Information
For this assay, SNP(s) [rs547721510] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PIP5KL1
Gene Name
phosphatidylinositol-4-phosphate 5-kinase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135219.1 Intron NP_001128691.1
NM_173492.1 Intron NP_775763.1
XM_017014279.1 Intron XP_016869768.1
Gene
ST6GALNAC4
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4
There are no transcripts associated with this gene.

View Full Product Details