Product Details

SNP ID
rs55944344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128884885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTTATTGACCTACCTCCCATGA[C/T]GGCGGAGACCATCTTCCATGTCTGC
Phenotype
MIM: 600547 MIM: 608360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KYAT1 PubMed Links

Gene Details

Gene
KYAT1
Gene Name
kynurenine aminotransferase 1
There are no transcripts associated with this gene.

Gene
LRRC8A
Gene Name
leucine rich repeat containing 8 family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127244.1 929 Intron NP_001120716.1
NM_001127245.1 929 Intron NP_001120717.1
NM_019594.3 929 Intron NP_062540.2
XM_005252095.2 929 Intron XP_005252152.1
XM_005252096.3 929 UTR 5 XP_005252153.1
XM_006717186.2 929 Intron XP_006717249.1
XM_011518863.1 929 Intron XP_011517165.1
XM_011518864.2 929 Intron XP_011517166.1
XM_011518865.2 929 Intron XP_011517167.1

View Full Product Details