Product Details

SNP ID
rs56044090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46333465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGAGCGTTTCCGCCAGCTGCAC[A/C]TACGAAAGCAGGTGTCTTACAGGTA
Phenotype
MIM: 601441
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DGKZ PubMed Links

Gene Details

Gene
DGKZ
Gene Name
diacylglycerol kinase zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105540.1 561 Intron NP_001099010.1
NM_001199266.1 561 Intron NP_001186195.1
NM_001199267.1 561 Intron NP_001186196.1
NM_001199268.1 561 Intron NP_001186197.1
NM_003646.3 561 Intron NP_003637.2
NM_201532.2 561 Missense Mutation ATA,CTA I64L NP_963290.1
NM_201533.3 561 Intron NP_963291.2
XM_005253181.3 561 Missense Mutation ATA,CTA I64L XP_005253238.1
XM_005253182.3 561 Intron XP_005253239.1
XM_006718354.1 561 Intron XP_006718417.1
XM_006718355.2 561 Intron XP_006718418.1
XM_011520422.1 561 Intron XP_011518724.1
XM_011520423.1 561 Intron XP_011518725.1
XM_017018455.1 561 Intron XP_016873944.1
XM_017018456.1 561 Intron XP_016873945.1
XM_017018457.1 561 Intron XP_016873946.1

View Full Product Details