Product Details

SNP ID
rs61942923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132146127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATCTTTATCGCCCCGAAAAGAA[A/G]TCCTGTACCAGTTACCATCGCTTCC
Phenotype
MIM: 612819
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX51 PubMed Links

Gene Details

Gene
DDX51
Gene Name
DEAD-box helicase 51
There are no transcripts associated with this gene.

Gene
NOC4L
Gene Name
nucleolar complex associated 4 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024078.2 Intron NP_076983.1
XM_017019960.1 Intron XP_016875449.1

View Full Product Details