Product Details

SNP ID
rs61917079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6199885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCGAAGGTTTGCAAGGAGACAGA[C/T]GAGGGCGAAATTAAGCCAGGCGGCT
Phenotype
MIM: 143030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD9 PubMed Links

Gene Details

Gene
CD9
Gene Name
CD9 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001769.3 42 Intron NP_001760.1
XM_005253814.3 42 Intron XP_005253871.1
XM_011521040.1 42 UTR 5 XP_011519342.1

View Full Product Details