Product Details

SNP ID
rs61944368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111362752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGCCATAGCACTCGTGCAGCC[G/T]GGCGAAGGGGGCCATGTCCAGGGGC
Phenotype
MIM: 614239
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM109A PubMed Links

Gene Details

Gene
FAM109A
Gene Name
family with sequence similarity 109 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177996.1 1067 Silent Mutation NP_001171467.1
NM_001177997.1 1067 Silent Mutation NP_001171468.1
NM_144671.4 1067 Silent Mutation NP_653272.2
XM_006719257.3 1067 Missense Mutation XP_006719320.1
XM_011537976.2 1067 Missense Mutation XP_011536278.1
XM_017018883.1 1067 Missense Mutation XP_016874372.1

View Full Product Details