Product Details

SNP ID
rs62027238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75674544 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCTGAGGCCAGGCCGGAGGGCC[A/G]ACCCCAGGGGCCCTCTGTATGCAAG
Phenotype
MIM: 601172
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSPG4 PubMed Links

Gene Details

Gene
CSPG4
Gene Name
chondroitin sulfate proteoglycan 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001897.4 8068 UTR 3 NP_001888.2

View Full Product Details