Product Details

SNP ID
rs62030195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:76217189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAACAGGAAGCCCAATTTTTCT[C/T]GTCTGGGTGAAGAGTACTGTGTATT
Phenotype
MIM: 608053
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ETFA PubMed Links

Gene Details

Gene
ETFA
Gene Name
electron transfer flavoprotein alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000126.3 Intron NP_000117.1
NM_001127716.1 Intron NP_001121188.1

View Full Product Details