Product Details

SNP ID
rs57274537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:79285779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAGCACAGTGCCAGCCAAGCAA[A/G]ACATGACTGTGGGCTACATCGGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD34C PubMed Links

Gene Details

Gene
ANKRD34C
Gene Name
ankyrin repeat domain 34C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146341.1 Intron NP_001139813.1
Gene
ANKRD34C-AS1
Gene Name
ANKRD34C antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details