Product Details

SNP ID
rs59521744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90966926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATAAACTTTTCATGTATATAAG[A/T]CAAAACCAAGTCTCCTAGGACCACT
Phenotype
MIM: 603484
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PRC1 PubMed Links

Gene Details

Gene
PRC1
Gene Name
protein regulator of cytokinesis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267580.1 1968 UTR 3 NP_001254509.1
NM_003981.3 1968 UTR 3 NP_003972.1
NM_199413.2 1968 UTR 3 NP_955445.1
XM_005254987.2 1968 UTR 3 XP_005255044.1
XM_006720759.1 1968 UTR 3 XP_006720822.1
XM_006720760.1 1968 Intron XP_006720823.1
XM_011522187.1 1968 Intron XP_011520489.1
XM_011522188.2 1968 Intron XP_011520490.1
XM_011522189.1 1968 Intron XP_011520491.1
XM_011522190.2 1968 Intron XP_011520492.1
XM_011522191.2 1968 Intron XP_011520493.1
XM_011522192.1 1968 Intron XP_011520494.1
XM_017022712.1 1968 UTR 3 XP_016878201.1
XM_017022713.1 1968 UTR 3 XP_016878202.1
XM_017022714.1 1968 Intron XP_016878203.1
XM_017022715.1 1968 UTR 3 XP_016878204.1
XM_017022716.1 1968 UTR 3 XP_016878205.1
XM_017022717.1 1968 UTR 3 XP_016878206.1
Gene
PRC1-AS1
Gene Name
PRC1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
RCCD1
Gene Name
RCC1 domain containing 1
There are no transcripts associated with this gene.

View Full Product Details