Product Details

SNP ID
rs62057951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67171449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCAGCAAAGGCTCCGAGGTGCC[A/G]GAGGCATAGTGAGTTCTCCGATCCC
Phenotype
MIM: 609077 MIM: 602438 MIM: 605235
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXL8 PubMed Links

Gene Details

Gene
FBXL8
Gene Name
F-box and leucine rich repeat protein 8
There are no transcripts associated with this gene.

Gene
HSF4
Gene Name
heat shock transcription factor 4
There are no transcripts associated with this gene.

Gene
KIAA0895L
Gene Name
KIAA0895 like
There are no transcripts associated with this gene.

Gene
NOL3
Gene Name
nucleolar protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185057.2 376 Intron NP_001171986.1
NM_001276307.1 376 Intron NP_001263236.1
NM_001276309.1 376 Intron NP_001263238.1
NM_001276311.1 376 Intron NP_001263240.1
NM_001276312.1 376 UTR 5 NP_001263241.1
NM_001276319.1 376 UTR 5 NP_001263248.1
NM_003946.6 376 Intron NP_003937.1
XM_005256217.2 376 Intron XP_005256274.1
XM_005256219.3 376 Intron XP_005256276.1
XM_011523424.2 376 UTR 5 XP_011521726.1
XM_017023843.1 376 Intron XP_016879332.1

View Full Product Details