Product Details

SNP ID
rs57807714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89931644 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACACAGATTAACCCACTTTCAT[C/T]TCAAAAAGACCTCCTGTCATCGTGC
Phenotype
MIM: 602661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TUBB3 PubMed Links

Gene Details

Gene
TUBB3
Gene Name
tubulin beta 3 class III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197181.1 Intron NP_001184110.1
NM_006086.3 Intron NP_006077.2

View Full Product Details