Product Details

SNP ID
rs55871953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:90057537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACTTGGGGTTCAGATATGTATG[C/G]AGACAAAATTAATTAGAACAGGAGG
Phenotype
MIM: 609759 MIM: 615805
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRDM7 PubMed Links

Gene Details

Gene
PRDM7
Gene Name
PR domain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098173.1 2147 UTR 3 NP_001091643.1
XM_005256274.3 2147 UTR 3 XP_005256331.1
XM_011522829.1 2147 UTR 3 XP_011521131.1
XM_011522831.2 2147 Intron XP_011521133.1
XM_017022882.1 2147 Intron XP_016878371.1
XM_017022883.1 2147 Intron XP_016878372.1
XM_017022884.1 2147 Intron XP_016878373.1
Gene
URAHP
Gene Name
urate (hydroxyiso-) hydrolase, pseudogene
There are no transcripts associated with this gene.

View Full Product Details