Product Details

SNP ID
rs56382961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1703190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTACTTACTCAGTTGTAATCACA[A/G]TCCAGTCCTGCCTTCCTCAAGTTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR22HG PubMed Links

Gene Details

Gene
MIR22HG
Gene Name
MIR22 host gene
There are no transcripts associated with this gene.

Gene
TLCD2
Gene Name
TLC domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164407.1 5501 UTR 3 NP_001157879.1

View Full Product Details