Product Details

SNP ID
rs61749864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:19022064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCGCTCCAGGACCTCAATGAT[A/G]TCGCCACGGCGGAAGCTGAGCTGCG
Phenotype
MIM: 604330
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRAP PubMed Links

Gene Details

Gene
GRAP
Gene Name
GRB2-related adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006613.3 612 Silent Mutation GAC,GAT D183D NP_006604.1
XM_017024029.1 612 Silent Mutation GAC,GAT D183D XP_016879518.1
XM_017024030.1 612 Missense Mutation ACA,ATA T127I XP_016879519.1
Gene
SLC5A10
Gene Name
solute carrier family 5 member 10
There are no transcripts associated with this gene.

View Full Product Details