Product Details

SNP ID
rs60198467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:19385202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGTCAGCGTACTTGGCATAGGC[C/T]GTGTTGTTCTCAAAGTCCTCCAAGT
Phenotype
MIM: 614144 MIM: 602521 MIM: 600596
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B9D1 PubMed Links

Gene Details

Gene
B9D1
Gene Name
B9 domain containing 1
There are no transcripts associated with this gene.

Gene
MAPK7
Gene Name
mitogen-activated protein kinase 7
There are no transcripts associated with this gene.

Gene
MFAP4
Gene Name
microfibrillar associated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198695.1 481 Silent Mutation ACA,ACG T163T NP_001185624.1
NM_002404.2 481 Silent Mutation ACA,ACG T139T NP_002395.1

View Full Product Details