Product Details

SNP ID
rs62064340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8394287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAATTTCATCCTTTGAGAATAT[A/G]TCTGATACTTTGGAAATGCCCAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF222 PubMed Links

Gene Details

Gene
RNF222
Gene Name
ring finger protein 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146684.2 196 UTR 5 NP_001140156.1
XM_011523978.2 196 Intron XP_011522280.1
XM_011523979.2 196 UTR 5 XP_011522281.1
XM_011523980.2 196 Intron XP_011522282.1
XM_011523981.2 196 Intron XP_011522283.1

View Full Product Details