Product Details

SNP ID
rs63751166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44349551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCAGTTGCTGCCCCTTCCCAGA[A/G]GTGAGCGTGCCATCAGCCCAGTGGA
Phenotype
MIM: 138945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM171A2 PubMed Links

Gene Details

Gene
FAM171A2
Gene Name
family with sequence similarity 171 member A2
There are no transcripts associated with this gene.

Gene
GRN
Gene Name
granulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002087.3 520 Silent Mutation GAA,GAG E88E NP_002078.1
XM_005257253.1 520 Silent Mutation GAA,GAG E88E XP_005257310.1

View Full Product Details