Product Details
- SNP ID
-
rs62120961
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:41268337 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGAAATGGAGCAGCAGCAGGCCT[A/G]TCGTCCAGGTAGGAAAATACACATG
- Phenotype
-
MIM: 109135
MIM: 605800
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AXL
PubMed Links
Gene Details
- Gene
- AXL
- Gene Name
- AXL receptor tyrosine kinase
There are no transcripts associated with this gene.
- Gene
- HNRNPUL1
- Gene Name
- heterogeneous nuclear ribonucleoprotein U like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001301016.2 |
740 |
Missense Mutation |
TAT,TGT |
Y48C |
NP_001287945.1 |
NM_001321208.1 |
740 |
Missense Mutation |
TAT,TGT |
Y37C |
NP_001308137.1 |
NM_001321211.1 |
740 |
Missense Mutation |
TAT,TGT |
Y37C |
NP_001308140.1 |
NM_007040.5 |
740 |
Missense Mutation |
TAT,TGT |
Y137C |
NP_008971.2 |
NM_144732.4 |
740 |
Missense Mutation |
TAT,TGT |
Y37C |
NP_653333.1 |
XM_005258459.3 |
740 |
Missense Mutation |
TAT,TGT |
Y137C |
XP_005258516.1 |
XM_005258461.4 |
740 |
Missense Mutation |
TAT,TGT |
Y48C |
XP_005258518.1 |
XM_005258463.3 |
740 |
Missense Mutation |
TAT,TGT |
Y37C |
XP_005258520.1 |
XM_005258464.3 |
740 |
Missense Mutation |
TAT,TGT |
Y37C |
XP_005258521.1 |
XM_005258465.4 |
740 |
Missense Mutation |
TAT,TGT |
Y37C |
XP_005258522.1 |
XM_011526392.2 |
740 |
Missense Mutation |
TAT,TGT |
Y137C |
XP_011524694.1 |
XM_011526393.2 |
740 |
Missense Mutation |
TAT,TGT |
Y37C |
XP_011524695.1 |
XM_011526394.2 |
740 |
Missense Mutation |
TAT,TGT |
Y37C |
XP_011524696.1 |
XM_011526395.2 |
740 |
Missense Mutation |
TAT,TGT |
Y37C |
XP_011524697.1 |
View Full Product Details