Product Details

SNP ID
rs62115999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51258360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGGAAGGATGTGAAGTCAAGAT[A/G]ATAAGCCTTTGATTCCTGGGCTGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIGLECL1 PubMed Links

Gene Details

Gene
SIGLECL1
Gene Name
SIGLEC family like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308437.1 Intron NP_001295366.1
NM_173635.2 Intron NP_775906.1
XM_005258799.4 Intron XP_005258856.1
XM_011526821.2 Intron XP_011525123.1
XM_017026647.1 Intron XP_016882136.1

View Full Product Details