Product Details

SNP ID
rs56334594
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51494254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATATCCAGAATATATTAAAAAAA[C/T]TCTTAGAACTCAACAGCAGAGAAAC
Phenotype
MIM: 606094
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CEACAM18 PubMed Links
Additional Information
For this assay, SNP(s) [rs57587487] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CEACAM18
Gene Name
carcinoembryonic antigen related cell adhesion molecule 18
There are no transcripts associated with this gene.

Gene
SIGLEC12
Gene Name
sialic acid binding Ig like lectin 12 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033329.2 Intron NP_201586.1
NM_053003.3 Intron NP_443729.1
XM_011527441.2 Intron XP_011525743.1
XM_011527442.2 Intron XP_011525744.1

View Full Product Details