Product Details

SNP ID
rs56142433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1877406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGATGAGCACGGGAGACGTGA[C/T]CTTGGACACCTTCTCGATGCTGCGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD17A PubMed Links

Gene Details

Gene
ABHD17A
Gene Name
abhydrolase domain containing 17A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130111.1 5262 Intron NP_001123583.1
NM_031213.3 5262 Intron NP_112490.3
XM_011528338.1 5262 Missense Mutation ATC,GTC I294V XP_011526640.1
XM_017027341.1 5262 Missense Mutation ATC,GTC I243V XP_016882830.1
XM_017027342.1 5262 Missense Mutation ATC,GTC I95V XP_016882831.1

View Full Product Details