Product Details
- SNP ID
-
rs62134283
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:680330 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGGCGTGCCGCATGCTGGGGGGC[C/G]GCCCGCGCTGCGAGTGCGCGCCCGA
- Phenotype
-
MIM: 605343
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FSTL3
PubMed Links
Gene Details
- Gene
- FSTL3
- Gene Name
- follistatin like 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005860.2 |
381 |
Missense Mutation |
CGC,GGC |
R116G |
NP_005851.1 |
- Gene
- PRSS57
- Gene Name
- protease, serine 57
There are no transcripts associated with this gene.
View Full Product Details