Product Details

SNP ID
rs62110694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12434869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGAACACTAACTCTAGAGACAG[A/C]AGATCAGACCTTATTTGCAAATTAT
Phenotype
MIM: 606697
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF443 PubMed Links

Gene Details

Gene
ZNF443
Gene Name
zinc finger protein 443
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005815.4 101 Intron NP_005806.2
XM_011527622.2 101 Intron XP_011525924.1
XM_011527623.2 101 Intron XP_011525925.2
XM_017026139.1 101 Intron XP_016881628.1
XM_017026140.1 101 UTR 5 XP_016881629.1

View Full Product Details