Product Details
- SNP ID
-
rs55726177
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:62940987 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATGGTAAATTGTGCCAAATAATC[A/G]TTGTACATTGAAGAATTCTGAATTC
- Phenotype
-
MIM: 604140
MIM: 611625
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DIDO1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10623609] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DIDO1
- Gene Name
- death inducer-obliterator 1
There are no transcripts associated with this gene.
- Gene
- GID8
- Gene Name
- GID complex subunit 8 homolog
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017896.2 |
|
Intron |
|
|
NP_060366.1 |
View Full Product Details