Product Details

SNP ID
rs62206635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63100362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCACGTGACATACCCTGCTAAG[A/C]GGGGCTTTGTTTTAACAGAATGGGG
Phenotype
MIM: 610556 MIM: 610557
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HAR1A PubMed Links

Gene Details

Gene
HAR1A
Gene Name
highly accelerated region 1A (non-protein coding)
There are no transcripts associated with this gene.

Gene
HAR1B
Gene Name
highly accelerated region 1B (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details