Product Details

SNP ID
rs61752459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:96884730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTCCCACGGGAACAAAGAGCCT[A/T]GAGGTATTGAAAATTCATGTCCTAG
Phenotype
MIM: 300108 MIM: 300767
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DIAPH2 PubMed Links

Gene Details

Gene
DIAPH2
Gene Name
diaphanous related formin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006729.4 823 Intron NP_006720.1
NM_007309.3 823 Intron NP_009293.1
Gene
RPA4
Gene Name
replication protein A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013347.4 823 Silent Mutation CTA,CTT L140L NP_037479.1

View Full Product Details