Product Details

SNP ID
rs56253937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:64190033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGGCCGGACCCTGGGCAGCTGA[A/G]GAATGCCATGGGTGATGCCCACAGG
Phenotype
MIM: 300647
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMER1 PubMed Links

Gene Details

Gene
AMER1
Gene Name
APC membrane recruitment protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152424.3 3527 Missense Mutation CCT,CTT P1085L NP_689637.3

View Full Product Details