Product Details
- SNP ID
-
rs56247447
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:99339589 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTAAAATCTACCTCTTTCCAGAG[C/T]GAAGCAGGTCAAATCCTTCATTTAT
- Phenotype
-
MIM: 103730
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADH1C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2903199,rs79904659] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADH1C
- Gene Name
- alcohol dehydrogenase 1C (class I), gamma polypeptide
View Full Product Details